Invitrogen
Manufacturer Code:PA569348
Catalog # PIPA569348
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the N terminal region of human SI |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: alpha-glucosidase; Isomaltase; maltase-glucoamylase (alpha-glucosidase); oligosaccharide alpha-1,6-glucosidase; pro-sucrase-isomaltase (EC 3.2.1.48-10); Sucrase; sucrase-isomaltase (alpha-glucosidase); Sucrase-isomaltase, intestinal
Gene Aliases: fd42f04; fd44a09; mgam; SI; SUCIMAL; wu:fd42f04; wu:fd44a09
UniProt ID: (Human) P14410, (Rabbit) P07768, (Rat) P23739
Entrez Gene ID: (Human) 6476, (Rabbit) 100009093, (Bovine) 504366, (Pig) 100623884, (Dog) 488141, (Horse) 100063242, (Guinea pig) 100731286, (Rat) 497756, (Zebrafish) 100148922
Molecular Function:
glucosidase
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.