Invitrogen
Manufacturer Code:PA561589
Catalog # PIPA561589
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human SHROOM3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: F-actin-binding protein; hShrmL; Protein Shroom3; Shroom-related protein; SHROOM3 SHROOM family member 3 APXL3 SHRM ShrmL F-actin-binding
Gene Aliases: APXL3; KIAA1481; MSTP013; SHRM; SHRML; SHROOM3
UniProt ID: (Human) Q8TF72
Entrez Gene ID: (Human) 57619
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
ion channel
ligand-gated ion channel
non-motor actin binding protein
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.