Novus Biologicals
Manufacturer Code:NBP281883
Catalog # NB017194
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Rabbit polyclonal SHROOM3 antibody was raised against an 18 amino acid peptide near the center of human SHROOM3. The immunogen is located within amino acids 850 - 900 of SHROOM3. Amino Acid Squence: CQLSGGASDSGRGPQRPDA |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: APXL3 MSTP013 SHRM ShrmL SHROOM3 shroom family member 3; F-actin-binding protein; hShrmL; Protein Shroom3; Shroom-related protein
Gene Aliases: APXL3; KIAA1481; MSTP013; SHRM; SHRML; SHROOM3
UniProt ID: (Human) Q8TF72
Entrez Gene ID: (Human) 57619
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
ion channel
ligand-gated ion channel
non-motor actin binding protein
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.