Invitrogen
Manufacturer Code:PA570281
Catalog # PIPA570281
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the N-terminal region of mouse SHQ1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: gene associated with retinoid and interferon-induced mortality 1; Protein SHQ1 homolog; SHQ1 homolog; SHQ1 Shq1p FLJ10539
Gene Aliases: 2810403P18Rik; GRIM-1; RGD1310610; SHQ1; Shq1p
UniProt ID: (Bovine) Q3MHH1, (Human) Q6PI26, (Mouse) Q7TMX5
Entrez Gene ID: (Bovine) 507486, (Dog) 476557, (Human) 55164, (Horse) 100062011, (Rabbit) 100352077, (Guinea pig) 100729827, (Mouse) 72171, (Rat) 297483
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.