Invitrogen
Manufacturer Code:PA534449
Catalog # PIPA534449
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 17 amino acid synthetic peptide near the amino terminus of human SHQ1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: gene associated with retinoid and interferon-induced mortality 1; Protein SHQ1 homolog; SHQ1 homolog; SHQ1 Shq1p FLJ10539
Gene Aliases: GRIM-1; SHQ1; Shq1p
UniProt ID: (Human) Q6PI26
Entrez Gene ID: (Human) 55164
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.