Novus Biologicals
Manufacturer Code:NBP242920
Catalog # NBP242920
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is raised against SHP-2/PTPN11. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BPTP3 EC 3.1.3.48 MGC14433 protein tyrosine phosphatase non-receptor type 11 protein tyrosine phosphatase-2 Protein-tyrosine phosphatase 1D Protein-tyrosine phosphatase 2C PTP-1D PTP-2C PTP2CNS1 Shp2 SHP-2 SHP2CFC SHPTP2 SH-PTP2Noonan syndrome 1 SH-PTP3 tyrosine-protein phosphatase non-receptor type 11; Protein-tyrosine phosphatase 1D; Protein-tyrosine phosphatase 2C; PTP-1D; PTP-2C; SH-PTP2; SH-PTP3; SHP-2; Tyrosine-protein phosphatase non-receptor type 11
Gene Aliases: BPTP3; CFC; JMML; METCDS; NS1; PTP-1D; PTP2C; PTPN11; SH-PTP2; SH-PTP3; SHP2; SHPTP2
UniProt ID: (Human) Q06124
Entrez Gene ID: (Human) 5781
Molecular Function: hydrolase phosphatase protein phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.