Invitrogen
Manufacturer Code:PA518692
Catalog # PIPA518692
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (KASRTSSKHKEE) corresponding to the internal amino acids of PTPN6 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: 70Z-SHP; hematopoietic cell phosphatase; Hematopoietic cell protein-tyrosine phosphatase; motheaten; Protein-tyrosine phosphatase 1C; Protein-tyrosine phosphatase SHP-1; PTP-1C; PTPN6 SHP1 protein tyrosine phosphatase non-receptor type 6 HCP HCPH HPTP1C PTP-1C SH-PTP1 SHP-1 SHP-1L 70 kda SHP-1L protein hematopoietic cell phosphatase hematopoietic cell protein-tyrosine phosphatase protein-tyrosine phosphatase 1C; PTPTY-42; SH-PTP1; SH2 phosphatase 1; SHP-1; Tyrosine-protein phosphatase non-receptor type 6
Gene Aliases: 70Z-SHP; HCP; HCPH; HPTP1C; me; PTP-1C; PTP1C; Ptph6; PTPN6; PTPTY-42; SH-PTP1; SHP-1; SHP-1L; SHP1
UniProt ID: (Human) P29350, (Rat) P81718, (Mouse) P29351
Entrez Gene ID: (Human) 5777, (Rat) 116689, (Mouse) 15170
Molecular Function:
hydrolase
phosphatase
protein phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.