Novus Biologicals
Manufacturer Code:NBP286805
Catalog # NB034427
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the N-terminal region of human SHOX2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Homeobox protein Og12X; Homeobox protein Og12X OG12SHOX homologous gene on chromosome 3 OG12Xshort stature homeobox protein 2 short stature homeobox 2 SHOTPaired-related homeobox protein SHOT; Paired-related homeobox protein SHOT; Short stature homeobox protein 2; SHOX homologous gene on chromosome 3
Gene Aliases: OG12; OG12X; SHOT; SHOX2
UniProt ID: (Human) O60902
Entrez Gene ID: (Human) 6474
Molecular Function: DNA binding protein helix-turn-helix transcription factor homeobox transcription factor nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.