Invitrogen
Manufacturer Code:PA5113508
Catalog # PIPA5113508
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Leucine-rich repeat protein SHOC-2 protein (3-89AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AU017287 KIAA0862 Leucine-rich repeat protein SHOC-2 mKIAA0862 Protein soc-2 homolog Protein sur-8 homolog Ras-binding protein SUR-8 RP11-348N5.2 Shoc2 SHOC2 leucine-rich repeat scaffold protein SHOC2 leucine rich repeat scaffold protein SIAA0862 SOC2 soc-2 (suppressor of clear) homolog soc-2 (suppressor of clear) homolog (C. elegans) soc-2 suppressor of clear homolog soc-2 suppressor of clear homolog (C. elegans) suppressor of clear C. elegans homolog of Sur8; Leucine-rich repeat protein SHOC-2; Protein soc-2 homolog; Protein sur-8 homolog; SHOC2 leucine-rich repeat scaffold protein; soc-2 suppressor of clear homolog
Gene Aliases: KIAA0862; SHOC2; SIAA0862; SOC2; SUR8
UniProt ID: (Human) Q9UQ13
Entrez Gene ID: (Human) 8036
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.