Invitrogen
Manufacturer Code:PA5106340
Catalog # PIPA5106340
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human Sur-8 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AU017287 KIAA0862 Leucine-rich repeat protein SHOC-2 mKIAA0862 Protein soc-2 homolog protein sur-8 homolog Ras-binding protein SUR-8 RP11-348N5.2 SHOC2 SHOC2 leucine-rich repeat scaffold protein SHOC2 leucine rich repeat scaffold protein SIAA0862 SOC2 soc-2 (suppressor of clear) homolog soc-2 (suppressor of clear) homolog (C. elegans) soc-2 suppressor of clear homolog soc-2 suppressor of clear homolog (C. elegans) suppressor of clear C. elegans homolog of Sur8; Leucine-rich repeat protein SHOC-2; Protein soc-2 homolog; Protein sur-8 homolog; Ras-binding protein SUR-8; SHOC2 leucine-rich repeat scaffold protein; soc-2 (suppressor of clear) homolog; soc-2 suppressor of clear homolog; suppressor of clear, C. elegans, homolog of
Gene Aliases: AU017287; KIAA0862; mKIAA0862; SHOC2; SIAA0862; SOC2; SUR8
UniProt ID: (Human) Q9UQ13, (Mouse) O88520, (Rat) Q6AYI5
Entrez Gene ID: (Human) 8036, (Mouse) 56392, (Rat) 309548
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.