Abnova
Manufacturer Code:PAB16760
Catalog # 89-118-597
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to N-terminus 21 amino acids of human SHOC2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C for three months. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of SHOC2.
For Research Use Only
Protein Aliases: FLJ60412; KIAA0862; Leucine-rich repeat protein SHOC-2; Protein soc-2 homolog; Protein sur-8 homolog; SHOC2 leucine-rich repeat scaffold protein; SIAA0862; SOC-2; soc-2 suppressor of clear homolog; SOC2; SUR-8; SUR8
Gene Aliases: KIAA0862; SHOC2; SIAA0862; SOC2; SUR8
UniProt ID: (Human) Q9UQ13
Entrez Gene ID: (Human) 8036
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.