Invitrogen
Manufacturer Code:PA543278
Catalog # PIPA543278
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human SHMT1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cytoplasmic serine hydroxymethyltransferase; cytosolic form; cytosolic serine hydroxymethyltransferase; Glycine hydroxymethyltransferase; serine hydroxymethyl transferase; serine hydroxymethyltransferase 1 (soluble); Serine hydroxymethyltransferase 1 (soluble) CSHMT MGC15229 MGC24556 SHMT; serine hydroxymethyltransferase 2 (mitochondrial); Serine hydroxymethyltransferase, cytosolic; Serine methylase; SHMT; zSHMT-1
Gene Aliases: AI324848; AI385541; C81125; CSHMT; LRRGT00032; MEL-32; mShmt; mshmt1; mshmt2; SHMT; SHMT1; SHMT2; zgc:66171; zgc:77524
UniProt ID: (Sheep) P35623, (Bovine) Q5E9P9, (Human) P34896, (Rabbit) P07511, (Mouse) P50431
Entrez Gene ID: (Sheep) 443544, (Horse) 100051522, (Bovine) 509002, (Dog) 489536, (Human) 6470, (Rabbit) 100009405, (Mouse) 20425, (Guinea pig) 100733584, (Rat) 287379, (Zebrafish) 394021
Molecular Function: methyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.