Invitrogen
Manufacturer Code:710410
Catalog # PI710410
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Oligoclonal |
Type | Antibody |
Clone | 23HCLC |
Immunogen | Peptide corresponding to amino acids 173184 of human SHH. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Recombinant Polyclonal Antibody
For Research Use Only
Protein Aliases: HHG-1; SHH; Shh N-terminal processed signaling domains; Shh unprocessed N-terminal signaling and C-terminal autoprocessing domains; ShhN; ShhNC; ShhNp; sonic hedgehog homolog; Sonic hedgehog protein; Sonic hedgehog protein N-product; TPT HHG1 HLP3 HPE3 SMMCI TPTPS MCOPCB5 SHH
Gene Aliases: HHG1; HLP3; HPE3; MCOPCB5; SHH; SMMCI; TPT; TPTPS; Vhh-1
UniProt ID: (Human) Q15465, (Rat) Q63673
Entrez Gene ID: (Human) 6469, (Rat) 29499
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.