Novus Biologicals
Manufacturer Code:NBP202434
Catalog # NBP202434
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3A1 |
Immunogen | Full length human recombinant protein of human SHC1 (NP_003020) produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: FLJ26504 p66 p66SHC SH2 domain protein C1 SHC SHC (Src homology 2 domain containing) transforming protein 1 SHC (Src homology 2 domain-containing) transforming protein 1 SHCA SHC-transforming protein 1 SHC-transforming protein 3 SHC-transforming protein A Src homology 2 domain-containing-transforming protein C1; SH2 domain protein C1; SHC (Src homology 2 domain containing) transforming protein 1; SHC (Src homology 2 domain-containing) transforming protein 1; SHC-transforming protein 1; SHC-transforming protein 3; SHC-transforming protein A; Src homology 2 domain-containing-transforming protein C1
Gene Aliases: SHC; SHC1; SHCA
UniProt ID: (Human) P29353
Entrez Gene ID: (Human) 6464
Molecular Function:
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.