Novus Biologicals
Manufacturer Code:NBP245825
Catalog # NBP245825
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1H10 |
Immunogen | Full length human recombinant protein of human SHBG(NP_001139751) produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ABP; ABPTestosterone-estrogen-binding globulin androgen binding protein MGC126834 MGC138391 SBP sex hormone-binding globulin Sex steroid-binding protein TeBG Testis-specific androgen-binding protein testosterone-binding beta-globulin Testosterone-estradiol-binding globulin; SBP; Sex hormone-binding globulin; Sex steroid-binding protein; SHBG; TeBG; Testis-specific androgen-binding protein; testosterone-binding beta-globulin; Testosterone-estradiol-binding globulin; Testosterone-estrogen-binding globulin
Gene Aliases: ABP; SBP; SHBG; TEBG
UniProt ID: (Human) P04278
Entrez Gene ID: (Human) 6462
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.