Invitrogen
Manufacturer Code:PA5101881
Catalog # PIPA5101881
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human SHB corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bA3J10.2 BC028832 SH2 domain containing adaptor protein B SH2 domain-containing adapter protein B Shb SHB (Src homology 2 domain containing) adaptor protein B SHB adaptor protein (a Src homology 2 protein) Src homology 2 domain containing adaptor protein B src homology 2 domain-containing transforming protein B; SH2 domain-containing adapter protein B; SHB (Src homology 2 domain containing) adaptor protein B; SHB adaptor protein (a Src homology 2 protein); Src homology 2 domain containing adaptor protein B
Gene Aliases: bA3J10.2; BC028832; SHB
UniProt ID: (Human) Q15464, (Mouse) Q6PD21
Entrez Gene ID: (Human) 6461, (Mouse) 230126
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.