Novus Biologicals
Manufacturer Code:NBP230557
Catalog # NBP230557
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to amino acids: TACGGKKLLNKCAASAAEESGAGKKDKVTIADDYSDPFDAKNDLKSKAGKGESAGYM |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bA3J10.2 RP11-3J10.8 SH2 domain-containing adapter protein B SHB (Src homology 2 domain containing) adaptor protein B SHB adaptor protein (a Src homology 2 protein) Src homology 2 domain containing adaptor protein B; SH2 domain-containing adapter protein B; SHB (Src homology 2 domain containing) adaptor protein B; SHB adaptor protein (a Src homology 2 protein); Src homology 2 domain containing adaptor protein B
Gene Aliases: bA3J10.2; SHB
UniProt ID: (Human) Q15464
Entrez Gene ID: (Human) 6461
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.