Proteintech
Manufacturer Code:146261AP150UL
Catalog # 50-554-693
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SHARPIN Fusion Protein Ag6201 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hSIPL1; hSIPL1 SHARPIN SIPL1; mSIPL1; protein kinase C-interacting protein RBCC like 1; SHANK-associated RH domain interacting protein; SHANK-associated RH domain interactor; Shank-associated RH domain-interacting protein; shank-interacting protein; Shank-interacting protein-like 1; Sharpin
Gene Aliases: 0610041B22Rik; AW121341; Conneck1; Cpdm; PSEC0216; RBCKL1; SHARPIN; SIPL1
UniProt ID: (Human) Q9H0F6, (Rat) Q9EQL9, (Mouse) Q91WA6
Entrez Gene ID: (Human) 81858, (Rat) 81859, (Mouse) 106025
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.