Novus Biologicals
Manufacturer Code:NBP147610C
Catalog # NBP147610C
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S69-46 |
Immunogen | Synthetic peptide amino acids 840-857 of rat Shank3 |
Conjugate | DyLight 650 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: DEL22q13.3 KIAA1650proline rich synapse associated protein 2 Proline-rich synapse-associated protein 2 ProSAP2 PSAP2SH3 and multiple ankyrin repeat domains protein 3 SH3 and multiple ankyrin repeat domains 3 shank postsynaptic density protein Shank3 SPANK-2; proline rich synapse associated protein 2; shank postsynaptic density protein
Gene Aliases: DEL22q13.3; PROSAP2; PSAP2; SCZD15; SPANK-2
Entrez Gene ID: (Human) 85358
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.