Invitrogen
Manufacturer Code:MA527634
Catalog # PIMA527634
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S23b-6 |
Immunogen | Fusion protein amino acids 84-309 (SH3/PDZ domains) of rat Shank2 (SH3 and multiple ankyrin repeat domains protein 2) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: cortactin SH3 domain-binding protein; Cortactin-binding protein 1; CortBP1; GKAP/SAPAP interacting protein; GKAP/SAPAP-interacting protein; proline rich synapse associated protein 1; proline-rich synapse associated protein 1; Proline-rich synapse-associated protein 1; ProSAP1; SH3 and multiple ankyrin repeat domains protein 2; SH3/ankyrin domain gene 2; Shank2; Shank2 PROSAP1 cortactin binding protein 1 KIAA1022 AUTS17 CortBP1 Proline-rich synapse-associated protein 1 SH3 and multiple ankyrin repeat domains protein 2 Cortactin-binding protein 1 proline-rich synapse associated protein 1 SPANK-3 GKAP/SAPAP interacting protein SHANK cortactin SH3 domain-binding protein ProSAP1 CORTBP1CTTNBP1 SH3 and multiple ankyrin repeat domains 2; SPANK-3
Gene Aliases: AUTS17; CORTBP1; CTTNBP1; KIAA1022; mKIAA1022; PROSAP1; SHANK; SHANK2; SPANK-3
UniProt ID: (Human) Q9UPX8, (Mouse) Q80Z38, (Rat) Q9QX74
Entrez Gene ID: (Human) 22941, (Mouse) 210274, (Rat) 171093
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.