R&D Systems
Manufacturer Code:AF7035
Catalog # AF7035
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human SHANK2 Lys849-Thr1029 Accession # Q9UPX8 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: AUTS17 cortactin binding protein 1 cortactin SH3 domain-binding protein Cortactin-binding protein 1 CortBP1 CORTBP1CTTNBP1 CTTNBP1 GKAP/SAPAP interacting protein KIAA1022 proline-rich synapse associated protein 1 Proline-rich synapse-associated protein 1 PROSAP1 SH3 and multiple ankyrin repeat domains 2 SH3 and multiple ankyrin repeat domains protein 2 SHANK Shank2 SPANK-3; cortactin SH3 domain-binding protein; Cortactin-binding protein 1; CortBP1; GKAP/SAPAP interacting protein; proline-rich synapse associated protein 1; Proline-rich synapse-associated protein 1; SH3 and multiple ankyrin repeat domains protein 2; Shank2
Gene Aliases: AUTS17; CORTBP1; CTTNBP1; KIAA1022; PROSAP1; SHANK; SHANK2; SPANK-3
UniProt ID: (Human) Q9UPX8
Entrez Gene ID: (Human) 22941
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.