Invitrogen
Manufacturer Code:MA527626
Catalog # PIMA527626
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S22-21 |
Immunogen | Fusion protein amino acids 469-691 (SH3/PDZ domains) of rat Shank1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: GKAP/SAPAP interacting protein; GKAP/SAPAP interacting protein OTTHUMP00000174437 SH3 and multiple ankyrin repeat domains 1 SH3 and multiple ankyrin repeat domains protein 1 SH3/ankyrin domain gene 1 SHAN1&lowbarHUMAN SHANK 1 Shank1 Shank1a Somatostatin receptor interacting protein Somatostatin receptor-interacting protein SPANK 1 SPANK1 SSTR interacting protein SSTR-interacting protein SSTRIP Synamon; GKAP/SAPAP-interacting protein; SAPAP-interacting protein synamon; SH3 and multiple ankyrin repeat domains 1; SH3 and multiple ankyrin repeat domains protein 1; SH3/ankyrin domain gene 1; Shank1; Somatostatin receptor-interacting protein; SPANK-1; SSTR-interacting protein; Synamon
Gene Aliases: SHANK1; Shank1a; SPANK-1; Spank1; SSTRIP; synamon
UniProt ID: (Human) Q9Y566, (Rat) Q9WV48, (Mouse) D3YZU1
Entrez Gene ID: (Human) 50944, (Rat) 78957, (Mouse) 243961
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.