Novus Biologicals
Manufacturer Code:NB300167SS
Catalog # NB018146
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | residues [SGPIYPGLFDIRSS] of the C-terminus of the SHANK1 protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: SH3 and multiple ankyrin repeat domains 1 SH3 and multiple ankyrin repeat domains protein 1 Shank1 Somatostatin receptor-interacting protein SSTR-interacting protein SSTRIPSPANK-1 synamon; SH3 and multiple ankyrin repeat domains protein 1; Shank1; Somatostatin receptor-interacting protein; SSTR-interacting protein
Gene Aliases: SHANK1; SPANK-1; SSTRIP; synamon
UniProt ID: (Human) Q9Y566
Entrez Gene ID: (Human) 50944
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.