Invitrogen
Manufacturer Code:PA557673
Catalog # PIPA557673
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human SH3PXD2B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Adapter protein HOFI; adaptor protein HOFI; Factor for adipocyte differentiation 49; SH3 and PX domain-containing protein 2B; Tyrosine kinase substrate with four SH3 domains
Gene Aliases: FAD49; FTHS; HOFI; KIAA1295; SH3PXD2B; TKS4; TSK4
UniProt ID: (Human) A1X283
Entrez Gene ID: (Human) 285590
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.