Invitrogen
Manufacturer Code:PA5117156
Catalog # PIPA5117156
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human SH3PXD2A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2310014D11Rik AA589508 Adapter protein TKS5 adaptor protein TKS5 AI256723 AI413738 C230050L11 EG329070 FISH Five SH3 domain-containing protein five SH3 domains Gm5098 KIAA0418 SH3 and PX domain-containing protein 2A SH3 and PX domains 2A SH3 multiple domains 1 SH3 multiple domains protein 1 SH3MD1 Sh3pxd2a Tks5 TSK5 Tyrosine kinase substrate with five SH3 domains; Adapter protein TKS5; adaptor protein TKS5; Five SH3 domain-containing protein; five SH3 domains; SH3 and PX domain-containing protein 2A; SH3 multiple domains 1; SH3 multiple domains protein 1; Tyrosine kinase substrate with five SH3 domains
Gene Aliases: 2310014D11Rik; AA589508; AI256723; AI413738; C230050L11; EG329070; FISH; Gm5098; KIAA0418; SH3MD1; SH3PXD2A; TKS5; TSK5
UniProt ID: (Human) Q5TCZ1, (Mouse) O89032
Entrez Gene ID: (Human) 9644, (Mouse) 14218
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.