Proteintech
Manufacturer Code:189761AP150UL
Catalog # 50-173-2299
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Adapter protein TKS5; Adapter protein TKS5 FISH KIAA0418 SH3 and PX domains 2A SH3 multiple domains protein 1 SH3MD1 SH3PXD2A TSK5; adaptor protein TKS5; Five SH3 domain-containing protein; five SH3 domains; SH3 and PX domain-containing protein 2A; SH3 multiple domains 1; SH3 multiple domains protein 1; Tyrosine kinase substrate with five SH3 domains
Gene Aliases: 2310014D11Rik; AA589508; AI256723; AI413738; C230050L11; EG329070; FISH; Gm5098; KIAA0418; SH3MD1; SH3PXD2A; TKS5; TSK5
UniProt ID: (Human) Q5TCZ1, (Mouse) O89032
Entrez Gene ID: (Human) 9644, (Rat) 309460, (Mouse) 14218
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.