Abnova
Manufacturer Code:H00009644K
Catalog # 89-206-396
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human SH3PXD2A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human SH3PXD2A peptide using ARM Technology.
For Research Use Only
Protein Aliases: Adapter protein TKS5; adaptor protein TKS5; FISH SH3MD1; Five SH3 domain-containing protein; SH3 and PX domain-containing protein 2A; SH3 multiple domains 1; SH3 multiple domains protein 1; Tyrosine kinase substrate with five SH3 domains
Gene Aliases: FISH; KIAA0418; SH3MD1; SH3PXD2A; TKS5
UniProt ID: (Human) Q5TCZ1
Entrez Gene ID: (Human) 9644
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.