Invitrogen
Manufacturer Code:PA557993
Catalog # PIPA557993
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human SH3BP4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BOG25 TTP EH-binding protein 10 SH3 domain-binding protein 4 transferrin receptor trafficking protein transferrin receptor-trafficking protein; EH-binding protein 10; SH3 domain-binding protein 4; SH3-domain binding protein 4; Transferrin receptor-trafficking protein
Gene Aliases: BOG25; EHB10; SH3BP4; TTP
UniProt ID: (Human) Q9P0V3
Entrez Gene ID: (Human) 23677
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.