Proteintech
Manufacturer Code:176911AP150UL
Catalog # 50-173-1723
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SH3BP4 Fusion Protein Ag11847 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BOG25 EH binding protein 10 EHB10 SH3 domain binding protein 4 SH3BP4 TTP; EH-binding protein 10; SH3 domain-binding protein 4; SH3-domain binding protein 4; Transferrin receptor-trafficking protein
Gene Aliases: AI594717; AW227605; BOG25; EHB10; SH3BP4; TTP
UniProt ID: (Human) Q9P0V3, (Mouse) Q921I6
Entrez Gene ID: (Human) 23677, (Rat) 64634, (Mouse) 98402
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.