Invitrogen
Manufacturer Code:PA530483
Catalog # PIPA530483
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein fragment corresponding to a region within amino acids 65 and 275 of Human SH3BP1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3BP-1; ARHGAP43 3BP-1 SH3 domain binding protein 1 SH3 domain-binding protein 1; SH3 domain-binding protein 1; SH3-domain binding protein 1
Gene Aliases: ARHGAP43; SH3BP1
UniProt ID: (Human) Q9Y3L3
Entrez Gene ID: (Human) 23616
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.