Proteintech
Manufacturer Code:205411AP150UL
Catalog # 50-173-2948
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SH3BP1 Fusion Protein Ag14575 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3BP 1 SH3 domain binding protein 1 SH3BP1; 3BP-1; SH3 domain-binding protein 1; SH3-domain binding protein 1
Gene Aliases: 3BP-1; 3bp1; ARHGAP43; SH3BP1
UniProt ID: (Human) Q9Y3L3, (Mouse) P55194
Entrez Gene ID: (Human) 23616, (Mouse) 20401
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.