Abnova
Manufacturer Code:H00023677A01
Catalog # 89-004-570
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | SH3BP4 (AAH57396 1 a.a. ~ 110 a.a) partial recombinant protein with GST tag. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant SH3BP4.
For Research Use Only
Protein Aliases: BOG25; EH-binding protein 10; SH3 domain-binding protein 4; SH3-domain binding protein 4; Transferrin receptor-trafficking protein; TTP
Gene Aliases: BOG25; EHB10; SH3BP4; TTP
UniProt ID: (Human) Q9P0V3
Entrez Gene ID: (Human) 23677
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.