Abnova
Manufacturer Code:PAB11521
Catalog # 89-115-152
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic phosphopeptide corresponding to residues surrounding Y127 of human SH2D1B. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised against synthetic phosphopeptide of SH2D1B.
For Research Use Only
Protein Aliases: EAT-2; EAT2; EWS/FLI1-activated transcript 2; SH2 domain-containing molecule EAT2; SH2 domain-containing protein 1B
Gene Aliases: EAT2; SH2D1B
UniProt ID: (Human) O14796
Entrez Gene ID: (Human) 117157
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.