Proteintech
Manufacturer Code:118711AP150UL
Catalog # 50-555-903
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SH2D1B Fusion Protein Ag2454 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EAT 2 EAT2 SH2 domain containing 1B SH2D1B; EAT-2; EWS/FLI1 activated transcript 2; EWS/FLI1-activated transcript 2; SH2 domain containing 1B; SH2 domain protein 1B1; SH2 domain-containing molecule EAT2; SH2 domain-containing protein 1B
Gene Aliases: EAT-2; EAT-2A; EAT2; Eat2a; RGD1563935; SH2D1B; Sh2d1b1
UniProt ID: (Human) O14796
Entrez Gene ID: (Human) 117157, (Rat) 501869, (Mouse) 26904
Molecular Function:
kinase
non-receptor tyrosine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.