Proteintech
Manufacturer Code:208251AP150UL
Catalog # 50-173-3109
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SGSM3 Fusion Protein Ag14800 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cx43-interacting protein of 85-kDa; MAP Merlin associated protein RabGAPLP RUSC3 RUTBC3 SGSM3; merlin binding protein; Merlin-associated protein; Rab-GTPase-activating protein-like protein; RabGAPLP; RUN and SH3 containing 3; RUN and TBC1 domain containing 3; RUN and TBC1 domain-containing protein 3; SGSM3; Small G protein signaling modulator 3; small G protein signaling modulator 3 protein
Gene Aliases: 1810012I01Rik; AI428557; BB175482; Cip85; MAP; R75178; RabGAP-5; RABGAP5; RABGAPLP; RUSC3; RUTBC3; SGSM3
UniProt ID: (Human) Q96HU1, (Mouse) Q8VCZ6
Entrez Gene ID: (Human) 27352, (Mouse) 105835
Molecular Function:
G-protein modulator
enzyme modulator
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.