Invitrogen
Manufacturer Code:PA572411
Catalog # PIPA572411
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | region 301-388/502 aa |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: heparan sulfate sulfamidase; heparan sulfate sulfatase; HSS MPS3A SFMD N-sulphoglucosamine sulphohydrolase heparan sulfate sulfatase mucopolysaccharidosis type IIIA sulfoglucosamine sulfamidase sulphamidase; mucopolysaccharidosis type IIIA; N-sulfoglucosamine sulfohydrolase; N-sulfoglucosamine sulfohydrolase (sulfamidase); N-sulphoglucosamine sulphohydrolase; Sulfoglucosamine sulfamidase; Sulphamidase
Gene Aliases: 4632406A19Rik; HSS; MPS3A; SFMD; SGSH
UniProt ID: (Human) P51688
Entrez Gene ID: (Human) 6448, (Dog) 403707, (Rat) 688293, (Mouse) 27029
Molecular Function: hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.