Invitrogen
Manufacturer Code:PA542767
Catalog # PIPA542767
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human SGPP2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: sphingosine 1-phosphate phosphohydrolase 2; Sphingosine-1-phosphatase 2; Sphingosine-1-phosphate phosphatase 2; Sphingosine-1-phosphate phosphatase 2 FLJ39004 SPP2; sphingosine-1-phosphate phosphotase 2; SPPase2
Gene Aliases: RGD1565739; SGPP2; SPP2; SPPase2
UniProt ID: (Human) Q8IWX5, (Mouse) Q810K3
Entrez Gene ID: (Human) 130367, (Horse) 100060934, (Rabbit) 100357833, (Guinea pig) 100731261, (Rat) 301543, (Mouse) 433323
Molecular Function:
hydrolase
phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.