Invitrogen
Manufacturer Code:PA535237
Catalog # PIPA535237
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 391-419 amino acids from the central region of human SGOL2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: SGOL2 Shugoshin-like 2 Shugoshin-2 Tripin; Shugoshin 2; Shugoshin-2; Shugoshin-like 2; Tripin
Gene Aliases: SGO2; SGOL2; TRIPIN
UniProt ID: (Human) Q562F6
Entrez Gene ID: (Human) 151246
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.