Novus Biologicals
Manufacturer Code:NB10060455
Catalog # NB10060455
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen recognized by this antibody maps to a region between residue 750 and 800 of human shugoshin-like 2 using the numbering given in entry NP_689737.3 (GeneID 151246). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ25211 MGC129665 Sgo2 Shugoshin-2 shugoshin-like 2 (S. pombe) Tripin TRIPINshugoshin-like 2; Shugoshin 2; Shugoshin-2; Shugoshin-like 2; Tripin
Gene Aliases: SGO2; SGOL2; TRIPIN
UniProt ID: (Human) Q562F6
Entrez Gene ID: (Human) 151246
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.