Invitrogen
Manufacturer Code:PA526744
Catalog # PIPA526744
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 338-365 amino acids from the C-terminal region of human SGMS2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Phosphatidylcholine:ceramide cholinephosphotransferase 2; SM synthase; SMS2 SM synthase phosphatidylcholine:ceramide cholinephosphotransferase 2; Sphingomyelin synthase 2
Gene Aliases: 4933405A16Rik; 5133401H06Rik; AI854299; SGMS2; SMS2
UniProt ID: (Human) Q8NHU3, (Mouse) Q9D4B1
Entrez Gene ID: (Human) 166929, (Mouse) 74442
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.