Invitrogen
Manufacturer Code:PA5113289
Catalog # PIPA5113289
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Phosphatidylcholine:ceramide cholinephosphotransferase 2 protein (1-79AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 4933405A16Rik 5133401H06Rik AI854299 MGC26963 Phosphatidylcholine:ceramide cholinephosphotransferase 2 Sgms2 SM synthase SMS2 Sphingomyelin synthase 2; Phosphatidylcholine:ceramide cholinephosphotransferase 2; SM synthase; Sphingomyelin synthase 2
Gene Aliases: RGD1305778; SGMS2; SMS2; spermatin
UniProt ID: (Human) Q8NHU3, (Rat) Q4JM44
Entrez Gene ID: (Human) 166929, (Rat) 310849
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.