Abnova
Manufacturer Code:PAB10380
Catalog # 89-114-822
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to human SGMS2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of SGMS2.
For Research Use Only
Protein Aliases: MGC26963; Phosphatidylcholine:ceramide cholinephosphotransferase 2; SM synthase; SMS2; Sphingomyelin synthase 2
Gene Aliases: SGMS2; SMS2
UniProt ID: (Human) Q8NHU3
Entrez Gene ID: (Human) 166929
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.