Abnova
Manufacturer Code:PAB10378
Catalog # 89-114-819
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to mouse Sgms1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of Sgms1.
For Research Use Only
Protein Aliases: 9530058O11Rik; AI841905; C80702; MGC30540; MOB; Phosphatidylcholine:ceramide cholinephosphotransferase 1; Protein Mob; SMS1; SMS1alpha1; SMS1alpha2; SMS1beta; SMS1gamma; Sor1; Sphingomyelin synthase 1; Tmem23; Transmembrane protein 23
Gene Aliases: 9530058O11Rik; AI841905; C80702; Mob; Sgms1; Sms1; Sor1; Tmem23
UniProt ID: (Mouse) Q8VCQ6
Entrez Gene ID: (Mouse) 208449
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.