Novus Biologicals
Manufacturer Code:NBP159886
Catalog # NBP159886
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to SLC5A4(solute carrier family 5 (low affinity glucose cotransporter) member 4) (NP_055042). The peptide sequence was selected from the N terminal of SLC5A4. Peptide sequence MASTVSPSTIAETPEPPPLSDHIRNAADISVIVIYFLVVMAV |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DJ90G24.4 low affinity sodium-glucose cotransporter Na(+)/glucose cotransporter 3 SAAT1low affinity sodium glucose cotransporter SGLT2 SGLT3 sodium transporter Sodium/glucose cotransporter 3 solute carrier family 5 (low affinity glucose cotransporter) member 4 solute carrier family 5 (neutral amino acid transporters system A) member 4 Solute carrier family 5 member 4; low affinity sodium glucose cotransporter; Na(+)/glucose cotransporter 3; sodium transporter; sodium/glucose cotransporter 3; solute carrier family 5 (glucose activated ion channel), member 4; solute carrier family 5 (low affinity glucose cotransporter), member 4; solute carrier family 5 (neutral amino acid transporters, system A), member 4; Solute carrier family 5 member 4
Gene Aliases: DJ90G24.4; SAAT1; SGLT3; SLC5A4
UniProt ID: (Human) Q9NY91
Entrez Gene ID: (Human) 6527
Molecular Function: carbohydrate transporter cation transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.