Proteintech
Manufacturer Code:243271AP150UL
Catalog # 50-173-4302
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SGLT3/SLC5A4 Fusion Protein Ag19391 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: low affinity sodium glucose cotransporter; low affinity sodium-glucose cotransporter; Na(+)/glucose cotransporter 3; Na(+)/glucose cotransporter 3 SAAT1 SGLT3 SLC5A4 Sodium/glucose cotransporter 3; sodium transporter; sodium/glucose cotransporter 3; solute carrier family 5 (glucose activated ion channel), member 4; solute carrier family 5 (low affinity glucose cotransporter), member 4; solute carrier family 5 (low affinity glucose cotransporter), member 4a; solute carrier family 5 (neutral amino acid transporters, system A), member 4; Solute carrier family 5 member 4; solute carrier family 5, member 4; solute carrier family 5, member 4a
Gene Aliases: DJ90G24.4; SAAT1; SGLT3; SLC5A4; Slc5a4a
UniProt ID: (Human) Q9NY91
Entrez Gene ID: (Human) 6527, (Rat) 294341
Molecular Function:
carbohydrate transporter
cation transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.