Invitrogen
Manufacturer Code:PA577461
Catalog # PIPA577461
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C)KDPAVGNISST(S)YQPR corresponding to amino acid residues 242-257 of rat SGLT2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: low affinity Na-dependent glucose transporter (SGLT2); low affinity sodium-dependent glucose cotransporter; Low affinity sodium-glucose cotransporter; Na(+)/glucose cotransporter 2; SGLT2 (extracellular); Sodium/glucose cotransporter 2; Solute carrier family 5 member 2
Gene Aliases: Sglt2; Slc5a2
UniProt ID: (Mouse) Q923I7, (Rat) P53792
Entrez Gene ID: (Mouse) 246787, (Rat) 64522
Molecular Function:
carbohydrate transporter
cation transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.