Novus Biologicals
Manufacturer Code:NBP192384
Catalog # NBP192384
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:FHEVGGYSGLFDKYLGAATSLTVSEDPAVGNISSFCYRPRPDSYHLL |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Low affinity sodium-glucose cotransporter; Low affinity sodium-glucose cotransporter Na(+)/glucose cotransporter 2 SGLT2sodium/glucose cotransporter 2 solute carrier family 5 (sodium/glucose cotransporter) member 2 solute carrier family 5 (sodium/glucose transporter) member 2 Solute carrier family 5 member 2; Na(+)/glucose cotransporter 2; Sodium/glucose cotransporter 2; solute carrier family 5 (sodium/glucose cotransporter), member 2; solute carrier family 5 (sodium/glucose transporter), member 2; Solute carrier family 5 member 2
Gene Aliases: SGLT2; SLC5A2
UniProt ID: (Human) P31639
Entrez Gene ID: (Human) 6524
Molecular Function:
carbohydrate transporter
cation transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.