Novus Biologicals
Manufacturer Code:NBP1458530.025MG
Catalog # NB018590
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide near the N-terminus of mouse SGLT-2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Low affinity sodium-glucose cotransporter; Low affinity sodium-glucose cotransporter Na(+)/glucose cotransporter 2 SGLT2sodium/glucose cotransporter 2 solute carrier family 5 (sodium/glucose cotransporter) member 2 solute carrier family 5 (sodium/glucose transporter) member 2 Solute carrier family 5 member 2; Na(+)/glucose cotransporter 2; Sodium/glucose cotransporter 2; solute carrier family 5 (sodium/glucose cotransporter), member 2; solute carrier family 5 (sodium/glucose transporter), member 2; Solute carrier family 5 member 2
Gene Aliases: SGLT2; SLC5A2
UniProt ID: (Human) P31639
Entrez Gene ID: (Human) 6524
Molecular Function:
carbohydrate transporter
cation transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.