Invitrogen
Manufacturer Code:PA569262
Catalog # PIPA569262
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human SGLT2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: glucose transporter; bSGLT2; low affinity Na-dependent glucose transporter (SGLT2); low affinity sodium-dependent glucose cotransporter; Low affinity sodium-glucose cotransporter; Na(+)/glucose cotransporter 2; Na(+)/nucleoside cotransporter; Na+/nucleoside cotransporter; SLC5A2 Na(+)/glucose cotransporter 2 SGLT2 Sodium-glucose transporter 2 Sodium/glucose cotransporter 2; sodium-glucose cotransporter-like 2; Sodium/glucose cotransporter 2; Sodium/nucleoside cotransporter; solute carrier family 5 (sodium/glucose cotransporter), member 2; solute carrier family 5 (sodium/glucose transporter), member 2; Solute carrier family 5 member 2
Gene Aliases: SGLT2; SLC5A2; SNST1
UniProt ID: (Human) P31639, (Rabbit) P26430, (Mouse) Q923I7, (Rat) P53792
Entrez Gene ID: (Human) 6524, (Dog) 492301, (Bovine) 399680, (Rabbit) 100009386, (Horse) 100064641, (Mouse) 246787, (Rat) 64522
Molecular Function:
carbohydrate transporter
cation transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.