Proteintech
Manufacturer Code:246541AP150UL
Catalog # 50-173-4473
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SGLT2 Fusion Protein Ag19163 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: low affinity sodium-dependent glucose cotransporter; Low affinity sodium-glucose cotransporter; Na(+)/glucose cotransporter 2; Na(+)/glucose cotransporter 2 SGLT2 SLC5A2 Sodium/glucose cotransporter 2; Sodium/glucose cotransporter 2; solute carrier family 5 (sodium/glucose cotransporter), member 2; solute carrier family 5 (sodium/glucose transporter), member 2; Solute carrier family 5 member 2
Gene Aliases: SGLT2; SLC5A2
UniProt ID: (Human) P31639, (Mouse) Q923I7
Entrez Gene ID: (Human) 6524, (Mouse) 246787
Molecular Function:
carbohydrate transporter
cation transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.